Learn More
Medchemexpress LLC 1H,5H-Imidazo[1,2-a]pyrrolo[1,2-d]pyrazin-5-one, 2,3,10,10a-tetrahydro-1-[(3-methyl-4-isoxazolyl)carbonyl]-10a-(6-methyl-3-pyridinyl)-, (10aS)- | 1323077-88-8 | 99.7% | 377.40 | 1 MG
SDP
Catalog No. 5000346157
Encompass_Preferred
Enzaplatovir (BTA-C585) is an orally bioavailable inhibitor for respiratory syncytial virus (RSV) infection and is described as a small-molecule RSV entry inhibitor. It is used for respiratory syncytial virus (RSV) infection and is relevant for RSV-associated lower respiratory tract infection (LRTI).
  • Orally bioavailable
  • Inhibitor for respiratory syncytial virus (RSV) infection
  • Small-molecule RSV entry inhibitor
  • Used for respiratory syncytial virus infection
  • Relevant for RSV-associated lower respiratory tract infection (LRTI)
Catalog No. 50-003-46157 Supplier Medchemexpress LLC Supplier No. HY109004A50MG
Enzaplatovir (BTA-C585) is an orally bioavailable inhibitor for respiratory syncytial virus (RSV) infection and is described as a small-molecule RSV entry inhibitor. It is used for respiratory syncytial virus (RSV) infection and is relevant for RSV-associated lower respiratory tract infection (LRTI).
  • Orally bioavailable
  • Inhibitor for respiratory syncytial virus (RSV) infection
  • Small-molecule RSV entry inhibitor
  • Used for respiratory syncytial virus infection
  • Relevant for RSV-associated lower respiratory tract infection (LRTI)