Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Benzimidazole, 2-(2-pyridinyl)- | 1137-68-4 | 99.9% | 195.22 | 10 G
SDP

Catalog No. 5000452494
Encompass_Preferred

2-(2′-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It functions as an anti-inflammatory agent and effectively inhibits Escherichia coli methionine aminopeptidase, making it suitable for biochemical research. This compound appears as a solid, ranging from pale purple to purple or colorless to light pink and is intended for research use only.

  • Functions as an anti-inflammatory agent
  • Inhibits Escherichia coli methionine aminopeptidase
  • IC50 of 13.3 μM in inhibiting human MetAP2 expressed in Sf9 cells
  • Molecular formula: C12H9N3
  • Molecular weight: 195.22
  • Purity: 99.88%
  • Ships at room temperature
  • Stable for up to 3 years at room temperature
  • Consistent with 1H NMR spectrum and LCMS

Catalog No. 50-004-52494 Supplier Medchemexpress LLC Supplier No. HYW00990610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-(2′-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It functions as an anti-inflammatory agent and effectively inhibits Escherichia coli methionine aminopeptidase, making it suitable for biochemical research. This compound appears as a solid, ranging from pale purple to purple or colorless to light pink and is intended for research use only.

  • Functions as an anti-inflammatory agent
  • Inhibits Escherichia coli methionine aminopeptidase
  • IC50 of 13.3 μM in inhibiting human MetAP2 expressed in Sf9 cells
  • Molecular formula: C12H9N3
  • Molecular weight: 195.22
  • Purity: 99.88%
  • Ships at room temperature
  • Stable for up to 3 years at room temperature
  • Consistent with 1H NMR spectrum and LCMS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.