Learn More
Medchemexpress LLC 1H-Benzimidazole, 2-(2-pyridinyl)- | 1137-68-4 | 99.9% | 195.22 | 100 G
SDP
Catalog No. 5000366389
Encompass_Preferred
2-(2'-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It can function as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand
  • Forms stable complexes with various transition metal ions
  • Can be used as an anti-inflammatory agent
  • Functions as an inhibitor of *Escherichia coli* methionine aminopeptidase
  • For research use only
Catalog No. 50-003-66389 Supplier Medchemexpress LLC Supplier No. HYW009906100G
2-(2'-Pyridyl)benzimidazole is a potentially tridentate ligand capable of forming stable complexes with various transition metal ions. It can function as an anti-inflammatory agent and an inhibitor of *Escherichia coli* methionine aminopeptidase.
  • Potentially tridentate ligand
  • Forms stable complexes with various transition metal ions
  • Can be used as an anti-inflammatory agent
  • Functions as an inhibitor of *Escherichia coli* methionine aminopeptidase
  • For research use only