Learn More
Medchemexpress LLC 1H-Benzimidazole-2-propanoic acid | 23249-97-0 | 99.7% | 250 MG
SDP
Catalog No. 5000413741
Encompass_Preferred
Procodazole is an orally active enhancer and a non-specific active immune protector against viral and bacterial infections. It also shows antiparasitic activity. Furthermore, Procodazole acts as a carbonic anhydrase inhibitor with antitumor activity.
  • Orally active enhancer.
  • Non-specific active immune protector against viral and bacterial infections.
  • Exhibits antiparasitic activity.
  • Carbonic anhydrase inhibitor with antitumor activity.
Catalog No. 50-004-13741 Supplier Medchemexpress LLC Supplier No. 250MGHYB1056
Procodazole is an orally active enhancer and a non-specific active immune protector against viral and bacterial infections. It also shows antiparasitic activity. Furthermore, Procodazole acts as a carbonic anhydrase inhibitor with antitumor activity.
  • Orally active enhancer.
  • Non-specific active immune protector against viral and bacterial infections.
  • Exhibits antiparasitic activity.
  • Carbonic anhydrase inhibitor with antitumor activity.