Learn More
Medchemexpress LLC 1H-Benzimidazole-5-carboxamide, 1-[(2E)-4-[5-(aminocarbonyl)-2-[[(1-ethyl-3-methyl-1H-pyrazol-5-yl)ca... | 2138299-33-7 | 99.9% | 849.94 | 50 MG
SDP
Catalog No. 5000436777
Encompass_Preferred
diABZI STING agonist-1 tautomerism is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130, 186 nM for human and mouse, respectively. It is a tautomerism of diABZI STING agonist-1.
  • Selective stimulator of interferon genes (STING) receptor agonist
  • EC50s of 130 nM for human and 186 nM for mouse
  • Shows high selectivity against over 350 kinases tested at 1 μM
  • Induces STING-dependent activation of type-I interferon and pro-inflammatory cytokines in vivo
  • Exhibits systemic exposure with a half-life of 1.4 h in vivo
  • Results in significant tumor growth inhibition and improved survival in vivo
Catalog No. 50-004-36777 Supplier Medchemexpress LLC Supplier No. HY112921A50MG
diABZI STING agonist-1 tautomerism is a selective stimulator of interferon genes (STING) receptor agonist, with EC50s of 130, 186 nM for human and mouse, respectively. It is a tautomerism of diABZI STING agonist-1.
  • Selective stimulator of interferon genes (STING) receptor agonist
  • EC50s of 130 nM for human and 186 nM for mouse
  • Shows high selectivity against over 350 kinases tested at 1 μM
  • Induces STING-dependent activation of type-I interferon and pro-inflammatory cytokines in vivo
  • Exhibits systemic exposure with a half-life of 1.4 h in vivo
  • Results in significant tumor growth inhibition and improved survival in vivo