Learn More
Medchemexpress LLC 1H-Benzimidazole, 6-bromo-4-fluoro-2-methyl-1-(1-methylethyl)- | 1231930-33-8 | 99.4% | C11H12BrFN2 | 10 G
SDP
Catalog No. 5000434856
Encompass_Preferred
Abemaciclib Impurity 1 is a drug intermediate used in the synthesis of various active compounds. It is supplied as an off-white to yellow solid for research use only.
  • High purity (99.4%)
  • Suitable for drug intermediate synthesis
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C
Catalog No. 50-004-34856 Supplier Medchemexpress LLC Supplier No. HY7806610G
Abemaciclib Impurity 1 is a drug intermediate used in the synthesis of various active compounds. It is supplied as an off-white to yellow solid for research use only.
  • High purity (99.4%)
  • Suitable for drug intermediate synthesis
  • Stable as powder for 3 years at -20°C
  • Stable as powder for 2 years at 4°C
  • Stable in solvent for 6 months at -80°C
  • Stable in solvent for 1 month at -20°C