Learn More
Medchemexpress LLC 1H-Benzimidazole, 6-methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]- | 119141-88-7 | 96.5% | 345.42 | 1 ML
SDP
Catalog No. 5000385914
Encompass_Preferred
Esomeprazole is a potent and orally active proton pump inhibitor that reduces acid secretion by inhibiting the H+, K+-ATPase in gastric parietal cells. It is used for symptomatic gastroesophageal reflux disease research.
  • Potent and orally active proton pump inhibitor
  • Reduces acid secretion by inhibiting H+, K+-ATPase
  • Used for symptomatic gastroesophageal reflux disease research
Catalog No. 50-003-85914 Supplier Medchemexpress LLC Supplier No. HY1702110MM/1ML
Esomeprazole is a potent and orally active proton pump inhibitor that reduces acid secretion by inhibiting the H+, K+-ATPase in gastric parietal cells. It is used for symptomatic gastroesophageal reflux disease research.
  • Potent and orally active proton pump inhibitor
  • Reduces acid secretion by inhibiting H+, K+-ATPase
  • Used for symptomatic gastroesophageal reflux disease research