Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1H-Benzo[D][1,2,3]Triazol-5-Ylboronic Acid Pinacol Ester

Catalog No. 715557250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-555-7250MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-555-7250MG Supplier Sigma Aldrich Supplier No. 715557250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1H-Benzo[D][1,2,3]Triazol-5-Ylboronic Acid Pinacol Ester

Specifications

CAS 163169-29-7
Melting Point 158°C to 162°C
Molecular Formula C28H28FeNOP
Linear Formula C28H28FeNOP
MDL Number MFCD15144869
Quantity 100 mg
Synonym (Sp)-1-(Diphenylphosphino)-2-[(4S)-4-isopropyl-2-oxazolyl]ferrocene
Molecular Weight (g/mol) 481.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.