Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium Chloride 98.0+%, TCI America™
SDP

Catalog No. B44865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44865G 5 g
B44861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44865G Supplier TCI America Supplier No. B44865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111198-09-5
Molecular Formula C25H21ClN3P
Molecular Weight (g/mol) 429.888
InChI Key YEEGDMCFIRPFRB-UHFFFAOYSA-M
PubChem CID 11988745
IUPAC Name benzotriazol-1-ylmethyl(triphenyl)phosphanium;chloride
SMILES C1=CC=C(C=C1)[P+](CN2C3=CC=CC=C3N=N2)(C4=CC=CC=C4)C5=CC=CC=C5.[Cl-]

Specifications

Melting Point 279°C
Color White
Quantity 5 g
Formula Weight 429.89
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material [(1H-Benzotriazol-1-yl)methyl]triphenylphosphonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.