Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-benzotriazole-4,5,6,7-d4 | 1185072-03-0 | MFCD09839986 | 99.9% | 123.15 | C6HD4N3 | 5mg
SDP

Catalog No. 5000269856
Encompass_Preferred

1H-Benzotriazole-4 5 6 7-d4 is the deuterium labeled 1H-Benzo[d][1 2 3]triazole[1]

Catalog No. 50-002-69856 Supplier Medchemexpress LLC Supplier No. HYY0688S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1H-Benzotriazole-4 5 6 7-d4 is the deuterium labeled 1H-Benzo[d][1 2 3]triazole[1]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.