Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Benzotriazole, ≥99.0%, Solstice

Catalog No. 60044756 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-756 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-756 Supplier Solstice Supplier No. 12798100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for amperometric det. of Cu,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-14-7
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD00005699
InChI Key QRUDEWIWKLJBPS-UHFFFAOYSA-N
Synonym 1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
PubChem CID 7220
ChEBI CHEBI:75331
IUPAC Name 2H-benzotriazole
SMILES C1=CC2=NNN=C2C=C1

Specifications

Melting Point 95°C to 99°C
Color Beige
pH 6.0 to 7.0 (at 100g/L 20°C)
Flash Point 170°C (338°F)
Ignition Residue ≤0.1%
Packaging Poly bottle
Quantity 100 g
UN Number NONH for all modes of transport
Beilstein 112133
Formula Weight 119.12g/mol
Percent Purity ≥99.0%
Physical Form Solid
Chemical Name or Material 1H-Benzotriazole
Show More Show Less
Hazard Statement P273-P305 + P351 + P338
Precautionary Statement H302 + H332-H319-H412
EINECSNumber 202-394-1
RTECSNumber DM1225000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.