Learn More
Medchemexpress LLC 1H-benzotriazolium, 1-[bis(dimethylamino)methylene]-, 3-oxide, hexafluorophosphate(1-) (1:1) | 94790-37-1 | MFCD00075445 | >98.0% | 379.25 | C11H16F6N5OP | 50g
SDP
Catalog No. 5000265936
Encompass_Preferred
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]
Catalog No. 50-002-65936 Supplier Medchemexpress LLC Supplier No. HYY091250G
Add to Cart
Add to Cart
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]