Learn More
Medchemexpress LLC 1H-benzotriazolium, 1-[bis(dimethylamino)methylene]-, 3-oxide, hexafluorophosphate(1-) (1:1) | 94790-37-1 | MFCD00075445 | 99.8% | 379.24 g/mol | C11H16F6N5OP | 100g
SDP
Catalog No. 5000269780
Encompass_Preferred
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]
Catalog No. 50-002-69780 Supplier Medchemexpress LLC Supplier No. HYY0912100G
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]