Learn More
Medchemexpress LLC 1H-benzotriazolium, 1-[bis(dimethylamino)methylene]-, 3-oxide, hexafluorophosphate | 94790-37-1 | MFCD00075445 | 98.0% | 379.24 g·mol^-1 | C11H16F6N5OP | 500g
SDP
Catalog No. 5000268524
Encompass_Preferred
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]
Catalog No. 50-002-68524 Supplier Medchemexpress LLC Supplier No. HYY0912500G
HBTU is a coupling agent commonly used for the activation of free carboxylic acids during the solution and solid phase peptide synthesis[1]