Learn More
Medchemexpress LLC 1H-Imidazole, 1-(4-ethynyl-2-methoxyphenyl)-4-methyl- | 1093980-57-4 | 99.8% | C13H12N2O | 1 G
SDP
Catalog No. 5000358085
Encompass_Preferred
1H-Imidazole, 1-(4-ethynyl-2-methoxyphenyl)-4-methyl- is a chemical compound identified by CAS Number 1093980-57-4. With a chemical formula of C13H12N2O and a molecular weight of 212.25, it appears as a white to off-white solid. This product has a high purity of 99.8% as determined by LCMS and is intended for research use only.
  • High purity of 99.8% (LCMS)
  • White to off-white solid appearance
  • Identified by CAS 1093980-57-4
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-58085 Supplier Medchemexpress LLC Supplier No. HY508491G
1H-Imidazole, 1-(4-ethynyl-2-methoxyphenyl)-4-methyl- is a chemical compound identified by CAS Number 1093980-57-4. With a chemical formula of C13H12N2O and a molecular weight of 212.25, it appears as a white to off-white solid. This product has a high purity of 99.8% as determined by LCMS and is intended for research use only.
  • High purity of 99.8% (LCMS)
  • White to off-white solid appearance
  • Identified by CAS 1093980-57-4
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month