Learn More
Medchemexpress LLC 1H-Imidazole, 2-(1-methylethyl)- | 36947-68-9 | 110.16 | 25 G
Supplier: Medchemexpress LLC 25GHYW001201
This 2-Isopropylimidazole product from MedChemExpress is a high-purity chemical with a molecular weight of 110.16. It is provided as a white to off-white solid and is suitable for research applications. The product offers specific storage conditions to maintain its stability.
- High purity 99.76% (HPLC)
- Molecular weight of 110.16
- Available as a white to off-white solid
- Powder stable for 3 years at -20°C and 2 years at 4°C
- In solvent stable for 6 months at -80°C and 1 month at -20°C