Learn More
Medchemexpress LLC 1H-Imidazole-2-carbaldehyde | 10111-08-7 | MFCD00003544 | 96.09 | 10 G
SDP
Catalog No. 5000460247
Encompass_Preferred
1H-Imidazole-2-carbaldehyde is a biochemical reagent suitable for life science research as a biological material or organic compound. This compound acts as a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with potential applications in suppressing type-2 diabetes.
  • Can be used as a biological material or organic compound for life science related research.
  • Acts as a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor.
  • Has potential application to suppress type-2 diabetes.
  • Used in the preparation of tridentate Schiff-base carboxylate-containing ligands by condensation reaction with amino acids.
  • Involved in the study of imidazole-directed allylation of aldimines.
Catalog No. 50-004-60247 Supplier Medchemexpress LLC Supplier No. HYW00732110G
1H-Imidazole-2-carbaldehyde is a biochemical reagent suitable for life science research as a biological material or organic compound. This compound acts as a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor with potential applications in suppressing type-2 diabetes.
  • Can be used as a biological material or organic compound for life science related research.
  • Acts as a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor.
  • Has potential application to suppress type-2 diabetes.
  • Used in the preparation of tridentate Schiff-base carboxylate-containing ligands by condensation reaction with amino acids.
  • Involved in the study of imidazole-directed allylation of aldimines.