Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Imidazole-2-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC08901DE
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC08901DE 5 g Amber Glass Bottle
CC08901DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC08901DE Supplier Thermo Scientific Chemicals Supplier No. CC08901DE
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16042-25-4
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
InChI Key KYWMCFOWDYFYLV-UHFFFAOYSA-N
Synonym 2-imidazolecarboxylic acid, imidazole-2-carboxylic acid, unii-o9g6515vc1, 1h-imidazolecarboxylic acid, 2-imidazolecarboxylicacid, 2-imidazocarboxylic acid, 1h-imidazole-2-carboxylicacid, imidazolecarboxylic, acmc-1avrt, 2-carboxy-1h-imidazole
PubChem CID 574321
IUPAC Name 1H-imidazole-2-carboxylic acid
SMILES C1=CN=C(N1)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 112.09
Percent Purity 97%
Chemical Name or Material 1H-Imidazole-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.