Learn More
Medchemexpress LLC 1h-Imidazole-4-carboxylic acid | 1072-84-0 | 112.09 | 5 G
SDP
Catalog No. 5000445939
Encompass_Preferred
1H-Imidazole-4-carboxylic acid is an organic compound featuring an imidazole ring and a carboxyl group. It stabilizes specific forms of metal ions through coordination, influencing chemical reactions. This compound plays a role in the hydrolysis of phosphate esters catalyzed by lanthanide metals and can modify dendrimers to alter surface properties and catalytic performance.
  • Stabilizes specific forms of metal ions
  • Influences chemical reactions
  • Role in hydrolysis of phosphate esters catalyzed by lanthanide metals
  • Can modify dendrimers to change surface properties
  • Can modify dendrimers to change catalytic performance
Catalog No. 50-004-45939 Supplier Medchemexpress LLC Supplier No. HYW0076955G
1H-Imidazole-4-carboxylic acid is an organic compound featuring an imidazole ring and a carboxyl group. It stabilizes specific forms of metal ions through coordination, influencing chemical reactions. This compound plays a role in the hydrolysis of phosphate esters catalyzed by lanthanide metals and can modify dendrimers to alter surface properties and catalytic performance.
  • Stabilizes specific forms of metal ions
  • Influences chemical reactions
  • Role in hydrolysis of phosphate esters catalyzed by lanthanide metals
  • Can modify dendrimers to change surface properties
  • Can modify dendrimers to change catalytic performance