Learn More
Medchemexpress LLC 1H-Imidazole-5-carboxaldehyde | 3034-50-2 | 100.0% | C4H4N2O | 10 G
SDP
Catalog No. 5000433986
Encompass_Preferred
1H-Imidazole-5-carboxaldehyde is a chemical reagent utilized in organic synthesis reactions, including the preparation of other imidazole compounds. It also serves as a biochemical dye and fluorescent probe.
  • Used as a chemical reagent in organic synthesis
  • Can be utilized to prepare other imidazole compounds, such as imidazole pyrimidine and imidazolone
  • Serves as a biochemical dye
  • Functions as a fluorescent probe
Catalog No. 50-004-33986 Supplier Medchemexpress LLC Supplier No. HY5930310G
1H-Imidazole-5-carboxaldehyde is a chemical reagent utilized in organic synthesis reactions, including the preparation of other imidazole compounds. It also serves as a biochemical dye and fluorescent probe.
  • Used as a chemical reagent in organic synthesis
  • Can be utilized to prepare other imidazole compounds, such as imidazole pyrimidine and imidazolone
  • Serves as a biochemical dye
  • Functions as a fluorescent probe