Learn More
Medchemexpress LLC 1H-Imidazole-5-carboxaldehyde | 3034-50-2 | 99.9% | C4H4N2O | 25 G
SDP
Catalog No. 5000358649
Encompass_Preferred
1H-Imidazole-5-carboxaldehyde serves as a versatile chemical reagent for organic synthesis reactions, enabling the creation of various imidazole compounds such as imidazole pyrimidine and imidazolone. It also finds utility as biochemical dyes and fluorescent probes in research applications.
  • Used as a chemical reagent in organic synthesis reactions
  • Prepares other imidazole compounds like imidazole pyrimidine and imidazolone
  • Functions as biochemical dyes
  • Functions as fluorescent probes
Catalog No. 50-003-58649 Supplier Medchemexpress LLC Supplier No. 25GHY59303
1H-Imidazole-5-carboxaldehyde serves as a versatile chemical reagent for organic synthesis reactions, enabling the creation of various imidazole compounds such as imidazole pyrimidine and imidazolone. It also finds utility as biochemical dyes and fluorescent probes in research applications.
  • Used as a chemical reagent in organic synthesis reactions
  • Prepares other imidazole compounds like imidazole pyrimidine and imidazolone
  • Functions as biochemical dyes
  • Functions as fluorescent probes