Learn More
Medchemexpress LLC 1H-Imidazole-5-carboxylic acid, 4-[1-hydroxy-1-(methyl-d3)ethyl-2,2,2-d3]-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]met | 1185144-74-4 | 98.7% | 452.54 | 1 MG
SDP
Catalog No. 5000391234
Encompass_Preferred
Olmesartan-d6 is a chemical compound with a molecular formula of C24H20D6N6O3 and a molecular weight of 452.54. It appears as a white to off-white solid. This product has been tested and complies with the given specifications.
  • Purity: 98.68% (HPLC)
  • Isotopic enrichment: 99.67%
  • 1H NMR spectrum consistent with structure
  • MS consistent with structure
Catalog No. 50-003-91234 Supplier Medchemexpress LLC Supplier No. HY17004S11MG
Olmesartan-d6 is a chemical compound with a molecular formula of C24H20D6N6O3 and a molecular weight of 452.54. It appears as a white to off-white solid. This product has been tested and complies with the given specifications.
  • Purity: 98.68% (HPLC)
  • Isotopic enrichment: 99.67%
  • 1H NMR spectrum consistent with structure
  • MS consistent with structure