Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Imidazole-5-methanol, 2-butyl-4-chloro-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, potassium salt | 124750-99-8 | 100.0% | 461.00 | 50 MG
SDP

Catalog No. 5000386489
Encompass_Preferred

Losartan potassium is an angiotensin II receptor type 1 (AT1) antagonist, competing with the binding of angiotensin II to AT1 with an IC50 of 20 nM. This product is for research use only.

  • Angiotensin II receptor type 1 (AT1) antagonist
  • IC50 of 20 nM
  • Purity 99.98%
  • Appearance: white to off-white solid
  • Store at 4°C, sealed, away from moisture

Catalog No. 50-003-86489 Supplier Medchemexpress LLC Supplier No. HY17512AR50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Losartan potassium is an angiotensin II receptor type 1 (AT1) antagonist, competing with the binding of angiotensin II to AT1 with an IC50 of 20 nM. This product is for research use only.

  • Angiotensin II receptor type 1 (AT1) antagonist
  • IC50 of 20 nM
  • Purity 99.98%
  • Appearance: white to off-white solid
  • Store at 4°C, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.