Learn More
Medchemexpress LLC 1H-Imidazole (hydrochloride) | 1467-16-9 | 100.0% | 104.54 | 25 G
SDP
Catalog No. 5000429323
Encompass_Preferred
1H-Imidazole hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as a ligand, buffer, and catalyst, playing an important role in drug synthesis and natural product extraction. It is also widely used in industrial fields such as the manufacturing of plastics, coatings, textiles, and electronic materials. While the compound has no direct medical applications, it is crucial for chemical research and laboratory studies.
  • Used as a ligand, buffer, and catalyst.
  • Plays a significant role in drug synthesis and extraction of natural products.
  • Widely used in industrial applications (plastics, coatings, textiles, electronic materials).
  • Important for chemical research and laboratory studies.
  • High purity: 99.96%.
Catalog No. 50-004-29323 Supplier Medchemexpress LLC Supplier No. 25GHYER010
1H-Imidazole hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It serves as a ligand, buffer, and catalyst, playing an important role in drug synthesis and natural product extraction. It is also widely used in industrial fields such as the manufacturing of plastics, coatings, textiles, and electronic materials. While the compound has no direct medical applications, it is crucial for chemical research and laboratory studies.
  • Used as a ligand, buffer, and catalyst.
  • Plays a significant role in drug synthesis and extraction of natural products.
  • Widely used in industrial applications (plastics, coatings, textiles, electronic materials).
  • Important for chemical research and laboratory studies.
  • High purity: 99.96%.