Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazol-6-ylboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC431020010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC431020010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431020010 Supplier Thermo Scientific Chemicals Supplier No. 431020010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 885068-10-0 | C7H7BN2O2 | 161.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885068-10-0
Molecular Formula C7H7BN2O2
Molecular Weight (g/mol) 161.96
MDL Number MFCD04972419
InChI Key ZKNLCHWRWRYPGG-UHFFFAOYSA-N
Synonym 6-indazolyboronic acid, 1h-indazol-6-yl-6-boronic acid, 1h-indazole-6-boronic acid, indazole-6-boronic acid, 1h-indazol-6-yl boronic acid, 6-indazolboronic acid, boronic acid, 1h-indazol-6-yl, 1h-indazole-6-boronicacid, 6-indazoleboronicacid, zlchem 1076
PubChem CID 24728617
IUPAC Name 1H-indazol-6-ylboronic acid
SMILES OB(O)C1=CC=C2C=NNC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180.0°C to 184.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazol-6-ylboronic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.