Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AC430090010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC430090010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430090010 Supplier Thermo Scientific Chemicals Supplier No. 430090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 677306-38-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD03840644
InChI Key KGKZHHIUOZGUNP-UHFFFAOYSA-N
Synonym indazole-4-carboxylic acid, 1h-indazole-4-carboxylicacid, 4-indazolecarboxylic acid, 4-carboxy-1h-indazole, 2h-indazole-4-carboxylic acid, pubchem19651, acmc-1b3os, ksc352q1d, 1h-indazole-4-carboxylic acid, 4-1h indazole carboxylic acid
PubChem CID 21982323
IUPAC Name 1H-indazole-4-carboxylic acid
SMILES C1=CC(=C2C=NNC2=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 209.5°C to 211.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazole-4-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.