Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Indazole, 5-[(1-methylcyclopropyl)oxy]- | 1626355-74-5 | 97.3% | 188.23 | 1 G
SDP

Catalog No. 5000401410
Encompass_Preferred

5-(1-Methylcyclopropoxy)-1H-indazole is a chemical compound with a molecular formula of C11H12N2O and a molecular weight of 188.23. It typically appears as a light yellow to yellow solid and has a purity of 97.34% by HPLC. This product is intended for research use only and has not been fully validated for medical applications.

  • CAS number: 1626355-74-5
  • Molecular formula: C11H12N2O
  • Molecular weight: 188.23
  • Purity (HPLC): 97.34%
  • Appearance: Light yellow to yellow solid
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month
  • Analytical data: 1H NMR and LCMS consistent with structure

Catalog No. 50-004-01410 Supplier Medchemexpress LLC Supplier No. HY425131G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-(1-Methylcyclopropoxy)-1H-indazole is a chemical compound with a molecular formula of C11H12N2O and a molecular weight of 188.23. It typically appears as a light yellow to yellow solid and has a purity of 97.34% by HPLC. This product is intended for research use only and has not been fully validated for medical applications.

  • CAS number: 1626355-74-5
  • Molecular formula: C11H12N2O
  • Molecular weight: 188.23
  • Purity (HPLC): 97.34%
  • Appearance: Light yellow to yellow solid
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month
  • Analytical data: 1H NMR and LCMS consistent with structure

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.