Learn More
Medchemexpress LLC 1H-Indazole, 5-[(1-methylcyclopropyl)oxy]- | 1626355-74-5 | 97.3% | 188.23 | 500 MG
SDP
Catalog No. 5000404829
Encompass_Preferred
This MedChemExpress product is 5-(1-Methylcyclopropoxy)-1H-indazole, a chemical compound with a molecular weight of 188.23 and a purity of 97.34%. It appears as a light yellow to yellow solid.
  • Consistent with structure by 1H NMR spectrum
  • Consistent with structure by LCMS
  • Ideal for research use only
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month
Catalog No. 50-004-04829 Supplier Medchemexpress LLC Supplier No. HY42513500MG
This MedChemExpress product is 5-(1-Methylcyclopropoxy)-1H-indazole, a chemical compound with a molecular weight of 188.23 and a purity of 97.34%. It appears as a light yellow to yellow solid.
  • Consistent with structure by 1H NMR spectrum
  • Consistent with structure by LCMS
  • Ideal for research use only
  • Store powder at -20°C for up to 3 years or 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months or -20°C for up to 1 month