Learn More
Medchemexpress LLC 1H-inden-1-amine, 2,3-dihydro-N-2-propyn-1-yl-, (1R)-, methanesulfonate (1:1) | 161735-79-1 | 99.8% | C13H17NO3S | 10MG

Supplier: Medchemexpress LLC HY1460510MG
Rasagiline mesylate is the mesylate salt of rasagiline, a selective, irreversible monoamine oxidase B (MAO-B) inhibitor supplied for research and analytical use. The product is provided in solid and solution forms and is suitable as an analytical standard and biochemical assay reagent.
- High purity: 99.8%.
- CAS number 161735-79-1 confirmed.
- Molecular formula C13H17NO3S; molecular weight 267.34 g/mol.
- Available as solid and as a 10 mM solution in water.
- Intended for research and analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.