Learn More
Medchemexpress LLC 1H-Indene-1,3(2H)-dione, 2,2-dihydroxy- | 485-47-2 | 100.0 % | 100 G
SDP
Catalog No. 5000409729
Encompass_Preferred
Ninhydrin is a chromogenic analytical probe used for the quantification of amino acids and proteins. It reacts with primary and secondary amines, producing a blue or purple reaction product, diketohydrindylidene-diketohydrindamine. It is utilized in the amino acid analysis of proteins; most amino acids, excluding proline, can be hydrolyzed and react with ninhydrin, after which they are colorimetrically quantified following chromatographic separation.
  • Chromogenic analytical probe.
  • Reacts with primary and secondary amines.
  • Solid appearance, white to light yellow color.
  • Quantification of amino acids and proteins.
  • Useful in amino acid analysis of proteins.
Catalog No. 50-004-09729 Supplier Medchemexpress LLC Supplier No. HYD0908100G
Ninhydrin is a chromogenic analytical probe used for the quantification of amino acids and proteins. It reacts with primary and secondary amines, producing a blue or purple reaction product, diketohydrindylidene-diketohydrindamine. It is utilized in the amino acid analysis of proteins; most amino acids, excluding proline, can be hydrolyzed and react with ninhydrin, after which they are colorimetrically quantified following chromatographic separation.
  • Chromogenic analytical probe.
  • Reacts with primary and secondary amines.
  • Solid appearance, white to light yellow color.
  • Quantification of amino acids and proteins.
  • Useful in amino acid analysis of proteins.