Learn More
Medchemexpress LLC 1H-Indene-1-ethanol, octahydro-4-hydroxy-ß,7a-dimethyl-, (ßS,1R,3aR,4S,7aR)- | 64190-52-9 | 98.2% | C₁₃H₂₄O₂ | 250 MG
SDP
Catalog No. 5000358429
Encompass_Preferred
Inhoffen Lythgoe diol is an organic molecule frequently used in chiral selective reactions during organic synthesis. It functions as both a catalyst and a ligand, finding broad applications in creating nitrogen- and sulfur-containing natural products, pharmaceuticals, and materials science. It plays a critical role in asymmetric synthesis. While it lacks direct medical applications, it is significant in the production and research of numerous important chemicals.
  • Used in chiral selective reactions during organic synthesis
  • Functions as both a catalyst and a ligand
  • Applied in creating nitrogen- and sulfur-containing natural products, pharmaceuticals, and materials science
  • Plays a critical role in asymmetric synthesis
  • For research use only
Catalog No. 50-003-58429 Supplier Medchemexpress LLC Supplier No. 250MGHY75920
Inhoffen Lythgoe diol is an organic molecule frequently used in chiral selective reactions during organic synthesis. It functions as both a catalyst and a ligand, finding broad applications in creating nitrogen- and sulfur-containing natural products, pharmaceuticals, and materials science. It plays a critical role in asymmetric synthesis. While it lacks direct medical applications, it is significant in the production and research of numerous important chemicals.
  • Used in chiral selective reactions during organic synthesis
  • Functions as both a catalyst and a ligand
  • Applied in creating nitrogen- and sulfur-containing natural products, pharmaceuticals, and materials science
  • Plays a critical role in asymmetric synthesis
  • For research use only