Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H-Indene, 90+%, stab. with 0.01% 4-tert-butylcatechol

Catalog No. AAA12192AD
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12192AD 50 mL
AAA12192AK 250 mL
AAA12192AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12192AD Supplier Thermo Scientific Chemicals Supplier No. A12192AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 95-13-6 | C9H8 | 116.163 g/mol

1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indene is a building block that has been used in the synthesis of isatins. It was used in the synthesis of new C60 derivative, indene-C60 bisadduct. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2.

Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.

Notes
Store at 4°C. Store in dark. Incompatible with oxidizing agent, light, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-13-6
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00003777
InChI Key YBYIRNPNPLQARY-UHFFFAOYSA-N
Synonym indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy
PubChem CID 7219
ChEBI CHEBI:41921
IUPAC Name 1H-indene
SMILES C1C=CC2=CC=CC=C21

Specifications

Melting Point ∽-2°C
Density 0.996
Boiling Point 181°C to 182°C
Flash Point 58°C (136°F)
Assay Percent Range ≥90%
Refractive Index 1.57
Quantity 50 mL
Beilstein 635873
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,acetone,ethyl ether,and ether. Insoluble in water.
Formula Weight 116.16
Physical Form Liquid
Chemical Name or Material Indene
Show More Show Less
Hazard Category H226-H335-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 202-393-6
RTECSNumber NK8225000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.