Learn More
Medchemexpress LLC 1H-indol-2-yl-[4-[2-(4-methoxyphenoxy)ethyl]piperazin-1-yl]methanone | 1386162-69-1 | MFCD32174273 | 99.5% | 379.45 g/mol | C22H25N3O3 | 5 MG
SDP
Catalog No. 5000222368
Encompass_Preferred
ML417 is a selective, brain-penetrant D3 dopamine receptor agonist supplied as a high-purity research compound for in vitro and in vivo pharmacology. It is suitable for receptor pharmacology studies and can be prepared in solution or used as a solid for dosing, with solubility and storage guidance provided.
Catalog No. 50-002-22368 Supplier Medchemexpress LLC Supplier No. HY1363905MG
ML417 is a selective, brain-penetrant D3 dopamine receptor agonist supplied as a high-purity research compound for in vitro and in vivo pharmacology. It is suitable for receptor pharmacology studies and can be prepared in solution or used as a solid for dosing, with solubility and storage guidance provided.