Learn More
Medchemexpress LLC 1H-Indol-5-amine | 5192-03-0 | 132.17 | 10 G
SDP
Catalog No. 5000448697
Encompass_Preferred
1H-Indol-5-amine (5-Aminoindole) is an organic compound featuring an indole ring structure. This derivative is a valuable intermediate for synthesizing other active compounds, including 11-aryl-3H-indolo[5,4-b]naphthyridine derivatives. It also serves as a ligand for preparing AI-Sepharose hydrophobic charge induction chromatography media.
  • Organic compound with indole ring structure
  • Key intermediate for synthesis of active compounds
  • Can be used as a ligand for preparing AI-Sepharose chromatography media
  • Suitable for protein adsorption and elution
Catalog No. 50-004-48697 Supplier Medchemexpress LLC Supplier No. HYW00750910G
1H-Indol-5-amine (5-Aminoindole) is an organic compound featuring an indole ring structure. This derivative is a valuable intermediate for synthesizing other active compounds, including 11-aryl-3H-indolo[5,4-b]naphthyridine derivatives. It also serves as a ligand for preparing AI-Sepharose hydrophobic charge induction chromatography media.
  • Organic compound with indole ring structure
  • Key intermediate for synthesis of active compounds
  • Can be used as a ligand for preparing AI-Sepharose chromatography media
  • Suitable for protein adsorption and elution