Learn More
Medchemexpress LLC 1H-Indol-5-ol, 3-(2-aminoethyl)- | 50-67-9 | 99.9% | 5 MG
SDP
Catalog No. 5000411415
Encompass_Preferred
Serotonin is a monoamine neurotransmitter in the CNS and an endogenous 5-HT receptor agonist. Serotonin is also a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. For research use only. We do not sell to patients.
  • Monoamine neurotransmitter in the CNS
  • Endogenous 5-HT receptor agonist
  • Catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM
Catalog No. 50-004-11415 Supplier Medchemexpress LLC Supplier No. HYB1473A5MG
Serotonin is a monoamine neurotransmitter in the CNS and an endogenous 5-HT receptor agonist. Serotonin is also a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. For research use only. We do not sell to patients.
  • Monoamine neurotransmitter in the CNS
  • Endogenous 5-HT receptor agonist
  • Catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM