Learn More
Medchemexpress LLC 1H-indol-5-ol, 3-(2-aminoethyl)- | 50-67-9 | MFCD00055054 | 99.9% | 50 MG
SDP
Catalog No. 5000411896
Encompass_Preferred
Serotonin, also known as 5-Hydroxytryptamine (5-HT), is a monoamine neurotransmitter in the central nervous system that acts as an endogenous 5-HT receptor agonist. It also functions as a catechol O-methyltransferase (COMT) inhibitor, binding non-competitively to the catechol substrate site with a Kᵢ of 44 μM. Research indicates that 100 μM Serotonin reduces COMT reaction velocity and produces robust hypersensitivity in vivo.
  • Monoamine neurotransmitter
  • Endogenous 5-HT receptor agonist
  • Catechol O-methyltransferase (COMT) inhibitor
  • Reduces COMT reaction velocity
  • Causes robust hypersensitivity in vivo
Catalog No. 50-004-11896 Supplier Medchemexpress LLC Supplier No. HYB1473A50MG
Serotonin, also known as 5-Hydroxytryptamine (5-HT), is a monoamine neurotransmitter in the central nervous system that acts as an endogenous 5-HT receptor agonist. It also functions as a catechol O-methyltransferase (COMT) inhibitor, binding non-competitively to the catechol substrate site with a Kᵢ of 44 μM. Research indicates that 100 μM Serotonin reduces COMT reaction velocity and produces robust hypersensitivity in vivo.
  • Monoamine neurotransmitter
  • Endogenous 5-HT receptor agonist
  • Catechol O-methyltransferase (COMT) inhibitor
  • Reduces COMT reaction velocity
  • Causes robust hypersensitivity in vivo