Learn More
Medchemexpress LLC 1H-indol-5-ol, 3-[4-(3,6-dihydro-4-phenyl-1(2H)-pyridinyl)butyl] hydrochloride | 108050-82-4 | 99.0% | 382.93 g/mol | C23H27ClN2O | 100 MG
SDP
Catalog No. 5000243157
Encompass_Preferred
Roxindole hydrochloride is the hydrochloride salt of roxindole (EMD 38362), a research tool compound investigated as a dopamine autoreceptor (D2-like) agonist and a 5-HT1A agonist that also inhibits serotonin uptake. It is used in vitro and in vivo to study dopaminergic and serotonergic signaling and to model psychiatric disorder pharmacology.
  • High purity (99.0%) suitable for analytical and research use.
  • Molecular weight 382.93 g/mol for accurate dosing calculations.
  • Hydrochloride salt form to improve solubility in polar solvents.
  • Appropriate for in vitro and in vivo pharmacology studies.
  • Provided in milligram quantities to support small-scale experiments.
Catalog No. 50-002-43157 Supplier Medchemexpress LLC Supplier No. HY106100A100MG
Roxindole hydrochloride is the hydrochloride salt of roxindole (EMD 38362), a research tool compound investigated as a dopamine autoreceptor (D2-like) agonist and a 5-HT1A agonist that also inhibits serotonin uptake. It is used in vitro and in vivo to study dopaminergic and serotonergic signaling and to model psychiatric disorder pharmacology.
  • High purity (99.0%) suitable for analytical and research use.
  • Molecular weight 382.93 g/mol for accurate dosing calculations.
  • Hydrochloride salt form to improve solubility in polar solvents.
  • Appropriate for in vitro and in vivo pharmacology studies.
  • Provided in milligram quantities to support small-scale experiments.