Learn More
Medchemexpress LLC 1H-Indole, 2,3-bis(1H-indol-3-ylmethyl)- | 138250-72-3 | 375.47 | 100 MG
SDP
Catalog No. 5000417160
Encompass_Preferred
2,3-Bis(3-indolylmethyl)indole significantly suppresses RANKL-induced osteoclast formation, actin ring formation, and bone resorption in a concentration-dependent manner. This product is for research use only.
  • Significantly suppresses RANKL-induced osteoclast formation, actin ring formation, and bone resorption in a concentration-dependent manner.
  • For research use only.
Catalog No. 50-004-17160 Supplier Medchemexpress LLC Supplier No. HYN10117100MG
2,3-Bis(3-indolylmethyl)indole significantly suppresses RANKL-induced osteoclast formation, actin ring formation, and bone resorption in a concentration-dependent manner. This product is for research use only.
  • Significantly suppresses RANKL-induced osteoclast formation, actin ring formation, and bone resorption in a concentration-dependent manner.
  • For research use only.