Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole-2,3-dione, 1-(phenylmethyl)- | 1217-89-6 | MFCD00187663 | 96.4% | 237.26 g/mol | C15H11NO2 | 250 MG
SDP

Catalog No. 5000208667
Encompass_Preferred

Benzylisatin is a biologically potent derivative of isatin used as a building block in medicinal chemistry research. It is commonly used as a precursor to synthesize biologically active Schiff bases and metal complexes and is supplied for biochemical assay and compound screening applications. The compound has CAS number 1217-89-6, molecular formula C15H11NO2, molecular weight 237.26 g/mol, and an analyzed purity of 96.39% by HPLC.

  • Biologically potent isatin derivative for medicinal chemistry research
  • Used to prepare Schiff bases and metal complexes
  • Molecular formula C15H11NO2 and molecular weight 237.26 g/mol
  • CAS number 1217-89-6 for unambiguous identification
  • Analyzed purity 96.39% (HPLC)

Catalog No. 50-002-08667 Supplier Medchemexpress LLC Supplier No. 250MGHYW046786
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzylisatin is a biologically potent derivative of isatin used as a building block in medicinal chemistry research. It is commonly used as a precursor to synthesize biologically active Schiff bases and metal complexes and is supplied for biochemical assay and compound screening applications. The compound has CAS number 1217-89-6, molecular formula C15H11NO2, molecular weight 237.26 g/mol, and an analyzed purity of 96.39% by HPLC.

  • Biologically potent isatin derivative for medicinal chemistry research
  • Used to prepare Schiff bases and metal complexes
  • Molecular formula C15H11NO2 and molecular weight 237.26 g/mol
  • CAS number 1217-89-6 for unambiguous identification
  • Analyzed purity 96.39% (HPLC)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.