Learn More
Medchemexpress LLC 1H-indole-2,4,5,6,7-d5-3-propanoic acid, α-oxo- | 85163-57-1 | MFCD08235196 | 96.0% | 208.22 | C11H4D5NO3 | 1mg
SDP
Catalog No. 5000268443
Encompass_Preferred
Indole-3-pyruvic acid-d5 is deuterated labeled Indole-3-pyruvic acid Indole-3-pyruvic acid a keto analogue of tryptophan is an orally active AHR agonist Indole-3-pyruvic acid has antioxidant properties and can be used in the research of inflammation anxiety[1][2][3]
Catalog No. 50-002-68443 Supplier Medchemexpress LLC Supplier No. HYW7183001MG
Indole-3-pyruvic acid-d5 is deuterated labeled Indole-3-pyruvic acid Indole-3-pyruvic acid a keto analogue of tryptophan is an orally active AHR agonist Indole-3-pyruvic acid has antioxidant properties and can be used in the research of inflammation anxiety[1][2][3]