Learn More
Medchemexpress LLC 1H-Indole-2-carbonitrile, 6-hydroxy-1-(1H-pyrazol-4-ylmethyl)-5-[[4-[[6-(2,2,2-trifluor... | 1857417-10-7 | 98.2% | 566.60 | 1 MG
SDP
Catalog No. 5000400774
Encompass_Preferred
MI-538 is an inhibitor of the interaction between menin and MLL fusion proteins with an IC50 of 21 nM. It is for research use only and not sold to patients.
  • Inhibits the proliferation of MLL leukemia cells with a GI50 of 83 nM.
  • Shows selectivity towards MLL fusion protein transformed cells.
  • Binds to menin with a Kd of 6.5 nM.
  • Leads to strong down-regulation of Hoxa9 and Meis1 gene expression.
  • Reduces tumor volume in mice xenografts.
  • Improved exposure and Cmax in blood plasma.
  • Low clearance and a half-life of about 1.6 hours.
  • High oral bioavailability (~50%).
  • Soluble in DMSO.
  • Soluble in various in vivo protocols, including 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline.
Catalog No. 50-004-00774 Supplier Medchemexpress LLC Supplier No. HY198101MG
MI-538 is an inhibitor of the interaction between menin and MLL fusion proteins with an IC50 of 21 nM. It is for research use only and not sold to patients.
  • Inhibits the proliferation of MLL leukemia cells with a GI50 of 83 nM.
  • Shows selectivity towards MLL fusion protein transformed cells.
  • Binds to menin with a Kd of 6.5 nM.
  • Leads to strong down-regulation of Hoxa9 and Meis1 gene expression.
  • Reduces tumor volume in mice xenografts.
  • Improved exposure and Cmax in blood plasma.
  • Low clearance and a half-life of about 1.6 hours.
  • High oral bioavailability (~50%).
  • Soluble in DMSO.
  • Soluble in various in vivo protocols, including 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline.