Learn More
Medchemexpress LLC 1h-Indole, 3-(2-chloro-4-pyrimidinyl)-1-methyl- | 1032452-86-0 | 99.5% | C13H10ClN3 | 10 G
SDP
Catalog No. 5000434866
Encompass_Preferred
This product is 1H-Indole, 3-(2-chloro-4-pyrimidinyl)-1-methyl-. It has a confirmed purity of 99.47% via HPLC, meeting specified quality standards.
  • High purity of 99.47% (HPLC)
  • Recommended storage for powder: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in-solvent solution: -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-34866 Supplier Medchemexpress LLC Supplier No. HY7886610G
This product is 1H-Indole, 3-(2-chloro-4-pyrimidinyl)-1-methyl-. It has a confirmed purity of 99.47% via HPLC, meeting specified quality standards.
  • High purity of 99.47% (HPLC)
  • Recommended storage for powder: -20°C for 3 years or 4°C for 2 years
  • Recommended storage for in-solvent solution: -80°C for 6 months or -20°C for 1 month