Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole, 3-(4-pyridinyl)- | 7272-84-6 | 99.5% | 194.23 g/mol | C13H10N2 | 5 MG
SDP

Catalog No. 5000263803
Encompass_Preferred

3-(4-Pyridyl)indole (Rockout) is a small-molecule Rho-kinase (ROCK) inhibitor used in cell biology to modulate actin cytoskeleton dynamics. It is provided as a high-purity solid and as DMSO solutions for in vitro and cellular assays.

  • Rho-kinase inhibitor activity with reported IC50 ~25 μM.
  • High purity: 99.5%.
  • Molecular weight 194.23 g/mol.
  • Available as solid and pre-made DMSO solutions for convenience.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Powder storage: -20 °C; in solvent: -80 °C (6 months) or -20 °C (1 month).
  • Used to inhibit blebbing and dissolve actin stress fibers in wound healing assays.

Catalog No. 50-002-63803 Supplier Medchemexpress LLC Supplier No. HY1123625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(4-Pyridyl)indole (Rockout) is a small-molecule Rho-kinase (ROCK) inhibitor used in cell biology to modulate actin cytoskeleton dynamics. It is provided as a high-purity solid and as DMSO solutions for in vitro and cellular assays.

  • Rho-kinase inhibitor activity with reported IC50 ~25 μM.
  • High purity: 99.5%.
  • Molecular weight 194.23 g/mol.
  • Available as solid and pre-made DMSO solutions for convenience.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Powder storage: -20 °C; in solvent: -80 °C (6 months) or -20 °C (1 month).
  • Used to inhibit blebbing and dissolve actin stress fibers in wound healing assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.