Learn More
Medchemexpress LLC 1H-Indole-3-acetamide | 879-37-8 | MFCD00005641 | 100.0% | 174.20 g/mol | C10H10N2O | 10 G
SDP
Catalog No. 5000301047
Encompass_Preferred
Indole-3-acetamide is an endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied as a powder for research and analytical applications, the compound has formula C10H10N2O and molecular weight 174.20 g/mol. Manufacturer documentation provides storage, solubility, and batch-specific purity details.
Catalog No. 50-003-01047 Supplier Medchemexpress LLC Supplier No. HYW01678410G
Indole-3-acetamide is an endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied as a powder for research and analytical applications, the compound has formula C10H10N2O and molecular weight 174.20 g/mol. Manufacturer documentation provides storage, solubility, and batch-specific purity details.