Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole-3-acetamide | 879-37-8 | MFCD00005641 | 100.0% | 174.20 g/mol | C10H10N2O | 100 G
SDP

Catalog No. 5000301046
Encompass_Preferred

Indole-3-acetamide is an indole-derived endogenous metabolite used as a biosynthetic intermediate of indole-3-acetic acid (IAA). The material is provided for research and analytical applications and is available as a solid or as a DMSO solution. It is intended for laboratory use only and is not for human or clinical applications.

  • High purity (99.95%).
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • Available as solid sizes up to 100 g and as a 10 mM solution in DMSO.
  • Reported solubility: soluble in DMSO; limited solubility in water and PBS; preparation notes available.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-003-01046 Supplier Medchemexpress LLC Supplier No. HYW016784100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide is an indole-derived endogenous metabolite used as a biosynthetic intermediate of indole-3-acetic acid (IAA). The material is provided for research and analytical applications and is available as a solid or as a DMSO solution. It is intended for laboratory use only and is not for human or clinical applications.

  • High purity (99.95%).
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • Available as solid sizes up to 100 g and as a 10 mM solution in DMSO.
  • Reported solubility: soluble in DMSO; limited solubility in water and PBS; preparation notes available.
  • Supplied with datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.