Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole-3-acetamide | 879-37-8 | MFCD00005641 | 100.0% | 174.20 g·mol⁻¹ | C10H10N2O | 25 G
SDP

Catalog No. 5000303057
Encompass_Preferred

Indole-3-acetamide is an indole-derived endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied for research use, it is typically used in studies of auxin biosynthesis, plant physiology, and microbial metabolism.

  • High purity (99.95% assay).
  • Chemical formula C10H10N2O; molecular weight 174.20 g·mol⁻¹.
  • Appearance: off-white to light brown solid.
  • Solubility: DMSO ≈100 mg/mL; water ≈1.3 mg/mL (ultrasonication may be required).
  • Storage: powder -20°C long term, 4°C short term; in solution -80°C long term.
  • Available in multiple pack sizes, including 25 g.
  • Safety and analytical documentation available (SDS, COA, data sheet).

Catalog No. 50-003-03057 Supplier Medchemexpress LLC Supplier No. 25GHYW016784
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide is an indole-derived endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied for research use, it is typically used in studies of auxin biosynthesis, plant physiology, and microbial metabolism.

  • High purity (99.95% assay).
  • Chemical formula C10H10N2O; molecular weight 174.20 g·mol⁻¹.
  • Appearance: off-white to light brown solid.
  • Solubility: DMSO ≈100 mg/mL; water ≈1.3 mg/mL (ultrasonication may be required).
  • Storage: powder -20°C long term, 4°C short term; in solution -80°C long term.
  • Available in multiple pack sizes, including 25 g.
  • Safety and analytical documentation available (SDS, COA, data sheet).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.