Learn More
Medchemexpress LLC 1H-indole-3-acetamide | 879-37-8 | MFCD00005641 | 100.0% | 174.20 g/mol | C10H10N2O | 25 MG
SDP
Catalog No. 5000302895
Encompass_Preferred
Indole-3-acetamide is an endogenous metabolite and analytical standard used in research and analytical applications. It functions as an intermediate in the biosynthesis of indole-3-acetic acid and is supplied as a characterized solid (CAS 879-37-8, C10H10N2O, MW 174.20 g/mol).
  • Analytical standard for research and analytical applications.
  • Intermediate in the biosynthesis pathway of indole-3-acetic acid.
  • High purity suitable for HPLC and analytical assays.
  • Supplied as a solid in stabilized packaging for storage.
  • Documented molecular formula and molecular weight for reproducibility.
Catalog No. 50-003-02895 Supplier Medchemexpress LLC Supplier No. 25MGHYW016784R
Indole-3-acetamide is an endogenous metabolite and analytical standard used in research and analytical applications. It functions as an intermediate in the biosynthesis of indole-3-acetic acid and is supplied as a characterized solid (CAS 879-37-8, C10H10N2O, MW 174.20 g/mol).
  • Analytical standard for research and analytical applications.
  • Intermediate in the biosynthesis pathway of indole-3-acetic acid.
  • High purity suitable for HPLC and analytical assays.
  • Supplied as a solid in stabilized packaging for storage.
  • Documented molecular formula and molecular weight for reproducibility.