Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole-3-acetamide | 879-37-8 | MFCD00005641 | 100.0% | 174.20 g/mol | C10H10N2O | 500 MG
SDP

Catalog No. 5000300417
Encompass_Preferred

Indole-3-acetamide is an endogenous indole metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied as a high-purity research chemical, it is intended for studies of auxin biosynthesis, plant physiology, and microbial tryptophan metabolism. Provided as a powdered reagent for analytical and laboratory applications.

  • High purity (99.95%).
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • Available in multiple pack sizes, including 500 mg.
  • Stable when stored as powder at -20°C for long-term storage.
  • Soluble in DMSO and water for assay preparation.
  • Intended for research and analytical use only.

Catalog No. 50-003-00417 Supplier Medchemexpress LLC Supplier No. HYW016784500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide is an endogenous indole metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied as a high-purity research chemical, it is intended for studies of auxin biosynthesis, plant physiology, and microbial tryptophan metabolism. Provided as a powdered reagent for analytical and laboratory applications.

  • High purity (99.95%).
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • Available in multiple pack sizes, including 500 mg.
  • Stable when stored as powder at -20°C for long-term storage.
  • Soluble in DMSO and water for assay preparation.
  • Intended for research and analytical use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.