Learn More
Medchemexpress LLC 1H-Indole-3-carboxylic acid, methyl ester | 942-24-5 | 99.9% | C10H9NO2 | 25 G
SDP
Catalog No. 5000359168
Encompass_Preferred
Methyl indole-3-carboxylate is a Sorangium metabolite. It exhibits weak activity against Gram-positive Nocardia sp. with a MIC of 33.3 μg/mL. This product is for research use only.
  • Shows weak activity against Gram-positive Nocardia sp.
  • Does not show significant cytotoxicity against L929, PC-3, and KB3-1 cells with IC50 values > 56.8 μM.
  • Available in solid and solution forms.
  • Can be dissolved in DMSO for in vitro experiments.
  • Protocols for in vivo dissolution are provided using combinations of DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.
Catalog No. 50-003-59168 Supplier Medchemexpress LLC Supplier No. 25GHY79635
Methyl indole-3-carboxylate is a Sorangium metabolite. It exhibits weak activity against Gram-positive Nocardia sp. with a MIC of 33.3 μg/mL. This product is for research use only.
  • Shows weak activity against Gram-positive Nocardia sp.
  • Does not show significant cytotoxicity against L929, PC-3, and KB3-1 cells with IC50 values > 56.8 μM.
  • Available in solid and solution forms.
  • Can be dissolved in DMSO for in vitro experiments.
  • Protocols for in vivo dissolution are provided using combinations of DMSO, PEG300, Tween-80, Saline, SBE-β-CD, and Corn Oil.