Learn More
Medchemexpress LLC 1H-indole-3-ethanamine, 5-methoxy | 608-07-1 | MFCD00005662 | 99.9% | 190.25 g/mol | C11H14N2O | 5 G
SDP
Catalog No. 5000302523
Encompass_Preferred
5-Methoxytryptamine is a tryptamine derivative and a melatonin metabolite that acts as a nonselective serotonin (5-HT) receptor agonist. Supplied for research use only, it is provided as a light yellow to yellow solid with reported LCMS purity around 99.6-99.9% and a molecular weight of 190.25 g/mol.
  • Acts as a nonselective serotonin (5-HT) receptor agonist.
  • Suitable for neuropharmacology and receptor binding studies.
  • High reported purity supports analytical and biological assays.
  • Supplied as a solid suitable for formulation or solution preparation.
  • Characterized by molecular formula C11H14N2O and molecular weight 190.25 g/mol.
  • Provided with batch-specific certificate of analysis for traceability.
Catalog No. 50-003-02523 Supplier Medchemexpress LLC Supplier No. HYW0151695G
5-Methoxytryptamine is a tryptamine derivative and a melatonin metabolite that acts as a nonselective serotonin (5-HT) receptor agonist. Supplied for research use only, it is provided as a light yellow to yellow solid with reported LCMS purity around 99.6-99.9% and a molecular weight of 190.25 g/mol.
  • Acts as a nonselective serotonin (5-HT) receptor agonist.
  • Suitable for neuropharmacology and receptor binding studies.
  • High reported purity supports analytical and biological assays.
  • Supplied as a solid suitable for formulation or solution preparation.
  • Characterized by molecular formula C11H14N2O and molecular weight 190.25 g/mol.
  • Provided with batch-specific certificate of analysis for traceability.